Skip to Content
Merck
All Photos(1)

Key Documents

33842

Supelco

HT-2 Toxin-13C22 solution

25 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C22H32O8
CAS Number:
Molecular Weight:
446.32
EC Number:
UNSPSC Code:
85151701

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

availability

not available in USA

concentration

25 μg/mL in acetonitrile

application(s)

cleaning products
cosmetics
food and beverages
personal care

mass shift

M+22

storage temp.

−20°C

SMILES string

[13CH3][13CH]([13CH3])[13CH2][13C](=O)O[13C@H]1[13CH2][13C@@]2([13CH2]O[13C]([13CH3])=O)[13C@H](O[13C@@H]3[13C@H](O)[13C@@H](O)[13C]2([13CH3])[13C]34[13CH2]O4)[13CH]=[13C]1[13CH3]

General description

Certan Vial

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service