Skip to Content
Merck
All Photos(1)

Key Documents

33621

Supelco

Fumonisin B1-13C34 solution

~25 μg/mL in acetonitrile: water, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$2,100.00

$2,100.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$2,100.00

About This Item

Empirical Formula (Hill Notation):
13C34H59NO15
CAS Number:
Molecular Weight:
755.58
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

$2,100.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~25 μg/mL in acetonitrile: water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+34

storage temp.

−20°C

SMILES string

[13CH3][13CH2][13CH2][13CH2][13C@@H]([13CH3])[13C@@H](O[13C](=O)[13CH2][13C@@H]([13CH2][13C](O)=O)[13C](O)=O)[13C@H]([13CH2][13C@@H]([13CH3])[13CH2][13C@H](O)[13CH2][13CH2][13CH2][13CH2][13C@@H](O)[13CH2][13C@H](O)[13C@H]([13CH3])N)O[13C](=O)[13CH2][13C@@H]([13CH2][13C](O)=O)[13C](O)=O

InChI

1S/C34H59NO15/c1-5-6-9-20(3)32(50-31(44)17-23(34(47)48)15-29(41)42)27(49-30(43)16-22(33(45)46)14-28(39)40)13-19(2)12-24(36)10-7-8-11-25(37)18-26(38)21(4)35/h19-27,32,36-38H,5-18,35H2,1-4H3,(H,39,40)(H,41,42)(H,45,46)(H,47,48)/t19-,20+,21-,22+,23+,24+,25+,

InChI key

UVBUBMSSQKOIBE-HTPIOKJKSA-N

General description

Fumonisin B1-13C34 is the 13C isotope-labelled analog of mycotoxin, Fumonisin B1.

Application

Fumonisin B1-13C34 may be used as an analytical standard for determining the levels of fumonisin B1 (FB1) in anis seed samples using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥92.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Fumonsins and Beauvericin in Anis seed by liquid chromatography/tandem mass spectrometry
Gorica JLV, et al.
Conference on medicinal and aromatic plants of southeast european countries, 214-214 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service