Skip to Content
Merck
All Photos(1)

Key Documents

33657

Supelco

2,3,4,5-Tetrachloroaniline solution

100 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl4N
CAS Number:
Molecular Weight:
230.91
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Nc1cc(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6H3Cl4N/c7-2-1-3(11)5(9)6(10)4(2)8/h1H,11H2

InChI key

GBKZRUCVLTWAML-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W de Wolf et al.
The Science of the total environment, 109-110, 457-459 (1991-12-01)
The lethal body burden (LBB) of 2,3,4,5-tetrachloroaniline in guppy (Poecilia reticulata), exposed for up to 14 days, has been determined at different aqueous concentrations of the test compound. It was found that the LBB is not constant during these 14
W de Wolf et al.
The Science of the total environment, 109-110, 383-386 (1991-12-01)
In this study the elimination kinetics of several chlorinated anilines and a tetrachlorobenzene in guppy, Poecilia reticulata, have been determined. The elimination of all the chlorinated anilines in fish showed a bi-phasic pattern, whereas the elimination characteristics of the tetrachlorobenzene
Didem Okutman Tas et al.
Environmental science & technology, 42(9), 3234-3240 (2008-06-05)
The effect of nitrate reduction onthe reductive biotransformation of pentachloronitrobenzene (PCNB), an organochlorine fungicide, was assessed with a mixed fermentative/methanogenic culture enriched from a contaminated estuarine sediment. Glucose and methanol served asthe electron and carbon source. PCNB at an initial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service