Skip to Content
Merck
All Photos(2)

Key Documents

33436

Supelco

Isofenphos-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$324.00

$324.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$324.00

About This Item

Empirical Formula (Hill Notation):
C14H22NO4PS
CAS Number:
Molecular Weight:
331.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$324.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(NC(C)C)Oc1ccccc1C(=O)OC(C)C

InChI

1S/C14H22NO4PS/c1-10(2)15-20(21,17-5)19-13-9-7-6-8-12(13)14(16)18-11(3)4/h6-11H,1-5H3,(H,15,21)

InChI key

IXTOWLKEARFCCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isofenphos-methyl is a broad spectrum, organophosphorus insecticide,[1] used to control soil pests in grain and oil crops such as click beetles, mole cricket and grub.[2]

Application

Isofenphos-methyl may be used as a reference standard for the determination of the insecticide in pepper samples using gas chromatography (GC) coupled with mass spectrometry (MS) and tandem mass spectrometry (MS/MS), respectively.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The toxicokinetics of methyl-ISP in rats.
N Shi et al.
Journal of Tongji Medical University = Tong ji yi ke da xue xue bao, 8(2), 78-82 (1988-01-01)
Acute toxicity of isofenphos-methyl and triazophos to 3 kinds or fresh water fish.
Shaonan L.
Environmental Pollution and Control, 3(1), 005-005 (1998)
Analyses of selected non-authorized insecticides in peppers by gas chromatography/mass spectrometry and gas chromatography/tandem mass spectrometry.
Mezcua M, et al.
Food Chemistry, 112(1), 221-225 (2005)
H Q Wu et al.
Journal of Tongji Medical University = Tong ji yi ke da xue xue bao, 9(1), 58-64 (1989-01-01)
Methyl-ISP, a newly developed organophosphorous insecticide, is used in China to treat and protect plants from pest infestation. Our studies demonstrated that methyl-ISP is metabolized rapidly in rat and mouse. Its toxicity was low, no obvious accumulative toxicity, chronic toxicity
Enantioseparation and determination of isofenphos-methyl enantiomers in wheat, corn, peanut and soil with supercritical fluid chromatography/tandem mass spectrometric method.
Chen X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1015(1), 13-21 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service