Skip to Content
Merck
All Photos(1)

Key Documents

333417

Sigma-Aldrich

Ligroin

reagent grade

Synonym(s):

Mineral spirits, Petroleum spirits

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12190000
grade:
reagent grade
bp:
60-80 °C (lit.)
vapor pressure:
25.8 psi
Pricing and availability is not currently available.

grade

reagent grade

vapor density

>1 (vs air)

vapor pressure

25.8 psi

form

liquid

expl. lim.

8 %

refractive index

n20/D 1.376 (lit.)

bp

60-80 °C (lit.)

density

0.656 g/mL at 25 °C (lit.)

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of
Sujith Prasad W Kumarasinghe et al.
International journal of dermatology, 41(12), 877-880 (2002-12-21)
Many methods have been employed, with variable success, in the treatment of cutaneous myiasis caused by Chrysomya species. Experiment 1: to assess the larvicidal effect of mineral turpentine (MT) and the main ingredient of MT, low aromatic white spirits (LAWS)
A Löf et al.
Pharmacology & toxicology, 85(2), 92-97 (1999-09-17)
Petroleum products with low content of aromatics have been increasingly used during the past years. This study investigates tissue disposition of dearomatised white spirit. In addition, brain neurotransmitter concentrations were measured. Male rats were exposed by inhalation to 0, 400

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service