Skip to Content
Merck
All Photos(1)

Key Documents

300314

Sigma-Aldrich

Petroleum ether

anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
grade:
anhydrous
bp:
30-60 °C (lit.)
vapor pressure:
25.8 psi ( 55 °C)
28.66 psi ( 55 °C)
7.99 psi ( 20 °C)
8.56 psi ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
28.66 psi ( 55 °C)
7.99 psi ( 20 °C)
8.56 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Petroleum ether can be used as a solvent:
  • In the radical ring-opening polymerization to synthesize degradable poly(vinyl esters)[1].
  • To synthesize di-block copolymer micelles[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

varepsilon-Thionocaprolactone: an accessible monomer for preparation of degradable poly (vinyl esters) by radical ring-opening polymerization
Ivanchenko O, et al.
Open Journal of Polymer Chemistry, 12, 1931-1938 (2021)
Synthesis of stable AuAg bimetallic nanoparticles encapsulated by diblock copolymer micelles
Menezes W, et al.
Nanoscale, 4, 1658-1664 (2012)
varepsilon-Thionocaprolactone: an accessible monomer for preparation of degradable poly (vinyl esters) by radical ring-opening polymerization
Ivanchenko O, et al.
Open Journal of Polymer Chemistry null
Eagleson M.
Concise Encyclopedia Chemistry, 783-783 (1994)
Rapid chromatographic technique for preparative separations with moderate resolution.
Still WC, et al.
The Journal of Organic Chemistry, 43(14), 2923-2925 (1978)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service