Skip to Content
Merck
All Photos(2)

Key Documents

32991

Supelco

Lanoconazole

VETRANAL®, analytical standard

Synonym(s):

2-[4-(2-Chlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-yl-acetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$300.00

$300.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$300.00

About This Item

Empirical Formula (Hill Notation):
C14H10ClN3S2
CAS Number:
Molecular Weight:
319.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$300.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccccc1C2CS\C(S2)=C(\C#N)n3ccnc3

InChI

1S/C14H10ClN3S2/c15-11-4-2-1-3-10(11)13-8-19-14(20-13)12(7-16)18-6-5-17-9-18/h1-6,9,13H,8H2/b14-12+

InChI key

ZRTQSJFIDWNVJW-WYMLVPIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lanoconazole is a member of the imidazole family[1] and an antifungal agent,[2] which finds applications as an antidermatophytic drug.[3]

Application

Lanoconazole may be used as a reference standard in the determination of lanoconazole in pharmaceutical formulations using high-performance liquid chromatography coupled with an ultraviolet detector (HPLC-UV).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Issues in Life Sciences: Botany and Plant Biology Research: 2011 Edition (2012)
Y Niwano et al.
Arzneimittel-Forschung, 47(9), 1056-1060 (1997-10-29)
To answer the question why topically applied lanoconazole (CAS 101530-10-3, NND-318) is so highly effective in the treatment of dermatomycoses in both animal models and human patients, the antifungal activity of lanoconazole in infected sites was investigated. 1. Distribution of
Y Niwano et al.
The Japanese journal of antibiotics, 48(1), 150-154 (1995-01-01)
The therapeutic efficacy of ointment and cream preparations of lanoconazole in a guinea pig model of tinea corporis was compared on the basis of degrees of improvement in local symptoms and negative culture rates. When infected animals were treated once
Application of experimental design for the optimization of forced degradation and development of a validated stability-indicating LC method for luliconazole in bulk and cream formulation.
Sonawane S and Gide P
Arabian Journal of Chemistry, 9, S1428-S1434 (2016)
M A Ghannoum et al.
Medical mycology, 48(3), 491-497 (2010-04-08)
The in vivo efficacy of terbinafine was compared to lanoconazole and luliconazole in the topical treatment of dermatophytosis caused by Trichophyton mentagrophytes using a guinea pig model. Topical antifungal treatment commenced three days post-infection, and each agent was applied once

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service