Skip to Content
Merck
All Photos(1)

Key Documents

C2162700

Ciclopirox olamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone ethanolammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2 · C2H7NO
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ciclopirox

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NCCO.CC1=CC(=O)N(O)C(=C1)C2CCCCC2

InChI

1S/C12H17NO2.C2H7NO/c1-9-7-11(13(15)12(14)8-9)10-5-3-2-4-6-10;3-1-2-4/h7-8,10,15H,2-6H2,1H3;4H,1-3H2

InChI key

MBRHNTMUYWQHMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclopirox olamine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Chouhan et al.
International journal of pharmaceutics, 436(1-2), 179-182 (2012-06-19)
Drug delivery by topical route for the treatment of onychomycosis, a nail fungal infection, is challenging due to the unique barrier properties of the nail plate which imparts high resistance to the passage of antifungal drugs. Permeation enhancers are used
Sae Hee Ko et al.
PloS one, 6(11), e27844-e27844 (2011-11-30)
Diabetic wounds remain a major medical challenge with often disappointing outcomes despite the best available care. An impaired response to tissue hypoxia and insufficient angiogenesis are major factors responsible for poor healing in diabetic wounds. Here we show that the
Aditya K Gupta et al.
The Journal of dermatological treatment, 25(3), 256-259 (2012-05-30)
Computerized nail planimetry is an objective means of accurately assessing affected nail areas, versus the standard visual estimation method. However, no published comparison of visual assessment with planimetry is currently available on onychomycosis to support this recommendation. This article evaluates
Siyuan Song et al.
Cancer research, 71(24), 7628-7639 (2011-10-20)
Excessive signaling from the Wnt pathway is associated with numerous human cancers. Using a high throughput screen designed to detect inhibitors of Wnt/β-catenin signaling, we identified a series of acyl hydrazones that act downstream of the β-catenin destruction complex to
Ciclopirox olamine: a hydroxypyridone antifungal agent.
B B Abrams et al.
Clinics in dermatology, 9(4), 471-477 (1991-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service