Skip to Content
Merck
All Photos(1)

Key Documents

32926

Supelco

Trichothecene Mix solution

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,140.00

$1,140.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$1,140.00

About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

$1,140.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

availability

not available in USA

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC) for all components
~10 ug/mL each in acetonitrile (3-AcDON, DON, NIV, FusX, HT-2, T-2, DAS, ZON)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service