Skip to Content
Merck
All Photos(1)

Documents

324825

Sigma-Aldrich

Basic Blue 41

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N4O6S2
CAS Number:
Molecular Weight:
482.57
Colour Index Number:
11105
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

form

powder

Quality Level

composition

Dye content, 40%

λmax

617 nm

SMILES string

COS([O-])(=O)=O.CCN(CCO)c1ccc(cc1)\N=N\c2sc3cc(OC)ccc3[n+]2C

InChI

1S/C19H23N4O2S.CH4O4S/c1-4-23(11-12-24)15-7-5-14(6-8-15)20-21-19-22(2)17-10-9-16(25-3)13-18(17)26-19;1-5-6(2,3)4/h5-10,13,24H,4,11-12H2,1-3H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

MHOFGBJTSNWTDT-UHFFFAOYSA-M

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service