Skip to Content
Merck
All Photos(2)

Key Documents

214566

Sigma-Aldrich

Acid Green 25

Dye content ≥60 %

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$204.00

About This Item

Empirical Formula (Hill Notation):
C28H20N2Na2O8S2
CAS Number:
Molecular Weight:
622.58
Colour Index Number:
61570
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

composition

Dye content, ≥60%

mp

235-238 °C (lit.)

λmax

608 nm
642 nm (2nd)

ε (extinction coefficient)

≥10000 at 604-612 nm
≥10200 at 640-646 nm
≥20000 at 281-287 nm
≥22700 at 251-257 nm
≥6400 at 407-413 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Cc1ccc(Nc2ccc(Nc3ccc(C)cc3S([O-])(=O)=O)c4C(=O)c5ccccc5C(=O)c24)c(c1)S([O-])(=O)=O

InChI

1S/C28H22N2O8S2.2Na/c1-15-7-9-19(23(13-15)39(33,34)35)29-21-11-12-22(30-20-10-8-16(2)14-24(20)40(36,37)38)26-25(21)27(31)17-5-3-4-6-18(17)28(26)32;;/h3-14,29-30H,1-2H3,(H,33,34,35)(H,36,37,38);;/q;2*+1/p-2

InChI key

FPAYXBWMYIMERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Acid green 25 is an anthraquinone, acid dye.

Application

Acid green 25 is commonly used in the textile industry to dye polyamide, wool, silk, and acrylic fibres.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W Hiebl et al.
Klinische Monatsblatter fur Augenheilkunde, 222(4), 309-311 (2005-04-22)
The azo dye trypan blue and the cyanine dye indocyanine green have been used in ophthalmology since the 1980 s to dye the anterior capsule in cataract operations and to stain epiretinal membranes in order to remove the membrana limitans
Xin-Hui Xu et al.
Molecules (Basel, Switzerland), 17(3), 2784-2795 (2012-03-08)
A new sorbent material, barium sulfate-Direct Blending Yellow D-3RNL hybrid (BSD), was synthesized and characterized by various methods. Both the anionic dyes, Reactive Brilliant Red X-3B and Weak Acid Green GS were hardly adsorbed by the BSD material, while the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service