Skip to Content
Merck
All Photos(2)

Key Documents

32104

Supelco

(±)-Naproxen-(methoxy-d3)

VETRANAL®, analytical standard

Synonym(s):

(RS)-2-[6-(Methoxy-d3)-2-naphthyl]propionic acid, 6-(Methoxy-d3)-α-methyl-2-naphthaleneacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$572.00

$572.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$572.00

About This Item

Empirical Formula (Hill Notation):
C14D3H11O3
CAS Number:
Molecular Weight:
233.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$572.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[2H]C([2H])([2H])Oc1ccc2cc(ccc2c1)C(C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/i2D3

InChI key

CMWTZPSULFXXJA-BMSJAHLVSA-N

General description

(±)-Naproxen-(methoxy-d3) may be used as a reference standard for the determination of (±)-naproxen-(methoxy-d3) in aqueous samples using solid-phase extraction (SPE) followed by ultra performance liquid chromatography-triple quadrupole mass spectrometry (UPLC-QqQ-MS/MS).[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pharmaceuticals in coastal systems using solid phase extraction (SPE) followed by ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).
Maria Baena-Nogueras R, et al.
Current Analytical Chemistry, 12(3), 183-201 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service