Skip to Content
Merck
All Photos(1)

Key Documents

31257

Supelco

1,2-Dibromo-3-chloropropane

PESTANAL®, analytical standard

Synonym(s):

DBCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCH2Cl
CAS Number:
Molecular Weight:
236.33
Beilstein:
1732077
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n/D 1.5542

bp

60-65 °C/7.5 mmHg

density

2.081 g/mL at 25 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClCC(Br)CBr

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dibromo-3-chloropropane (DCBP) is a brominated organochlorine nematocide used in agriculture as a soil fumigant.[1]

Application

DCBP may be used as a reference standard for the determination of the analyte in crops using gas-liquid chromatographic detection method.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 1B - Repr. 1A - STOT RE 2 Oral

Target Organs

Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic activity of 1, 2-dibromo-3-chloropropane, a widely-used fumigant.
Rosenkranz HS.
Bulletin of Environmental Contamination and Toxicology, 14(1), 8-12 (1975)
A method for the determination of 1, 3-dichloropropene, 1, 2-dibromoethane, and 1, 2-dibromo-3-chloropropane in food crops.
Newsome WH and Panopio LG
Journal of Agricultural and Food Chemistry, 25(5), 998-1000 (1977)
Unwelcome exposure.
Tim Rogers
Lancet (London, England), 364(9439), 1025-1026 (2004-09-24)
R Sass
International journal of health services : planning, administration, evaluation, 30(3), 491-514 (2000-12-08)
The poisoning of Costa Rican banana workers by multinational corporations' excessive use of pesticides is not a local issue; it is embedded in a dominant ideology expressed by the phenomenon of globalization. This ideology seeps into every aspect of our
1,2-Dibromo-3-chloropropane.
Report on carcinogens : carcinogen profiles, 12, 134-135 (2011-08-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service