Skip to Content
Merck
All Photos(1)

Key Documents

31233

Sigma-Aldrich

Iron(III) nitrate nonahydrate

puriss. p.a., ACS reagent, ≥98%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
solid

grade

ACS reagent
puriss. p.a.

Assay

≥98%

form

solid

mp

47 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Cd: ≤10 mg/kg
Fe2+: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermal decomposition of iron(III) nitrate nonahydrate has been reported. Fe2O3 was obtained as the major decomposition product at 523K.

Application

Iron(III) nitrate nonahydrate may be employed as an inorganic precursor for the preparation of iron(III)-doped titania nanoparticles by modified sol-gel method. Ferric nitrate nonahydrate supported on silica may be used for the preparation of benzils.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of iron (III)-doped titania nanoparticles and its application for photodegradation of sulforhodamine-B pollutant.
Kumbhar A and Chumanov G.
Journal of Nanoparticle Research, 7(4-5), 489-498 (2005)
Thermal decomposition of Ni (II) and Fe (III) nitrates and their mixture.
Elmasry MAA, et al.
Journal of Thermal Analysis and Calorimetry, 52(2), 489-495 (1998)
Sangchul Hwang et al.
Chemosphere, 78(5), 563-568 (2009-12-05)
Fenton-driven oxidation of methyl tert-butyl ether (MTBE) (0.11-0.16mM) in batch reactors containing ferric iron (5mM) and hydrogen peroxide (H(2)O(2)) (6mM) (pH=3) was performed to investigate MTBE transformation mechanisms. Independent variables included the forms of iron (Fe) (Fe(2)(SO(4))(3).9H(2)O and Fe(NO(3))(3).9H(2)O), H(2)O(2)
Guang-Li Wang et al.
Analytica chimica acta, 747, 92-98 (2012-09-19)
An innovative and versatile functional colorimetric sensor for melamine (MA) and H(2)O(2) was developed with simplicity, excellent selectivity and ultrasensitivity. The detection mechanism was based on the "oxidative etching-aggregation" of silver nanoparticles (AgNPs) by the cooperation effect of MA and
Jacob R Hascalovici et al.
Journal of neurochemistry, 108(1), 72-81 (2008-12-03)
Up-regulation of heme oxygenase-1 (HO-1) and altered cholesterol (CH) metabolism are characteristic of Alzheimer-diseased neural tissues. The liver X receptor (LXR) is a molecular sensor of CH homeostasis. In the current study, we determined the effects of HO-1 over-expression and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service