Skip to Content
Merck
All Photos(1)

Key Documents

31099

Supelco

Chlordimeform

PESTANAL®, analytical standard

Synonym(s):

N-(4-Chloro-2-methylphenyl)-N′,N′-dimethylformamidine, Chlorodimeform, Chlorophenamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2
CAS Number:
Molecular Weight:
196.68
Beilstein:
9256367
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CN(C)\C=N\c1ccc(Cl)cc1C

InChI

1S/C10H13ClN2/c1-8-6-9(11)4-5-10(8)12-7-13(2)3/h4-7H,1-3H3/b12-7+

InChI key

STUSTWKEFDQFFZ-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlordimeform is a formamidine insecticide/acaricide, effective against pests resistant to many organophosphates and chlorinated hydrocarbon insecticides.

Application

Chlordimeform may be used as a reference standard for the determination of the analyte:
  • In honey by a method based on solid-phase microextraction (SPME) followed by gas chromatography with microwave-induced plasma atomic emission detection.
  • In Chinese material medica (CMM), a food source supplement using modified QuEChERS (Quick Easy Cheap Effective Rugged Safe) sample preparation procedure and gas chromatography tandem mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multi-residue method for simultaneous determination of 74 pesticides in Chinese material medica using modified QuEChERS sample preparation procedure and gas chromatography tandem mass spectrometry.
Liu XQ, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1015(1), 1- 12 (2016)
Solid-phase microextraction and gas chromatography with atomic emission detection for multiresidue determination of pesticides in honey.
Campillo N, et al.
Analytica Chimica Acta, 562(1), 9- 15 (2006)
Inhibition of monoamine oxidase by the pesticide chlordimeform and related compounds.
Aziz SA, Knowles CO.
Nature, 242(5397), 417-418 (1973)
Solid-phase microextraction and gas chromatography with atomic emission detection for multiresidue determination of pesticides in honey.
Campillo, Natalia, et al.
Analytica Chimica Acta, 562.1, 9-15 (2006)
Biochemical and physiological effects of chlordimeform.
Matsumura F and Beeman RW
Environmental Health Perspectives, 14, 71-71 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service