Skip to Content
Merck
All Photos(1)

Key Documents

29650

Supelco

Cyclooctene

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14
CAS Number:
Molecular Weight:
110.20
Beilstein:
1901031
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.470

bp

32-34 °C/12 mmHg (lit.)

density

0.848 g/mL at 20 °C (lit.)

SMILES string

C1CCCC=CCC1

InChI

1S/C8H14/c1-2-4-6-8-7-5-3-1/h1-2H,3-8H2/b2-1-

InChI key

URYYVOIYTNXXBN-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service