Skip to Content
Merck
All Photos(1)

Key Documents

25800

Sigma-Aldrich

3-Chloroperbenzoic acid

technical, ~70% (RT)

Synonym(s):

3-Chloroperoxybenzoic acid, MCPBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO3H
CAS Number:
Molecular Weight:
172.57
Beilstein:
608317
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical

Assay

~70% (RT)

reaction suitability

reagent type: oxidant

impurities

~10% 3-chlorobenzoic acid
~20% water

mp

69-71 °C (lit.)

storage temp.

2-8°C

SMILES string

OOC(=O)c1cccc(Cl)c1

InChI

1S/C7H5ClO3/c8-6-3-1-2-5(4-6)7(9)11-10/h1-4,10H

InChI key

NHQDETIJWKXCTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the epoxidation of olefins

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Org. Perox. D - Skin Corr. 1C - Skin Sens. 1

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.M. Rubottom et al.
Organic Syntheses, 7, 282-282 (1990)
Tatyana Spolitak et al.
Archives of biochemistry and biophysics, 493(2), 184-191 (2009-11-03)
Our previous rapid-scanning stopped-flow studies of the reaction of substrate-free cytochrome P450cam with peracids [T. Spolitak, J.H. Dawson, D.P. Ballou, J. Biol. Chem. 280 (2005) 20300-20309; J. Inorg. Biochem. 100 (2006) 2034-2044; J. Biol. Inorg. Chem. 13 (2008) 599-611] spectrally
Evelyn B Rodriguez et al.
Journal of food science, 74(9), C674-C682 (2010-05-25)
To gain a better understanding of the reactions and the underlying mechanisms of the oxidative degradation of lycopene, the products formed by epoxidation with m-chloroperbenzoic acid (MCPBA), oxidative cleavage with KMnO(4), and autoxidation in low-moisture and aqueous model systems, under
Shin Kamijo et al.
Organic letters, 12(18), 4195-4197 (2010-08-26)
The direct oxidation of ether sp(3) C-H bonds using the new reagent system mCPBA/CCl(3)CN/MeCN has been developed. CCl(3)CN in MeCN drastically alters the reactivity of m-chloroperbenzoic acid (mCPBA), and chemoselective transformation of methyl ethers to ketones was realized under mild
Which oxidant is really responsible for P450 model oxygenation reactions? A kinetic approach.
Alicja Franke et al.
Angewandte Chemie (International ed. in English), 47(28), 5238-5242 (2008-06-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service