Skip to Content
Merck
All Photos(1)

Key Documents

234109

Sigma-Aldrich

Methyl Orange

certified by the BSC, certified by the Biological Stain Commission

Synonym(s):

4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid sodium salt, Acid Orange 52, Helianthin, Orange III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N3NaO3S
CAS Number:
Molecular Weight:
327.33
Colour Index Number:
13025
Beilstein:
4732884
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

certified by the Biological Stain Commission

Agency

certified by the BSC

composition

Dye content, ≥85%

pH

3.0-4.4, pink to yellow

λmax

507 nm

SMILES string

[Na+].CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S([O-])(=O)=O

InChI

1S/C14H15N3O3S.Na/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20;/h3-10H,1-2H3,(H,18,19,20);/q;+1/p-1/b16-15+;

InChI key

STZCRXQWRGQSJD-GEEYTBSJSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in the Wilson-Ezrin method for pituitary acidophils; Kreyberg′s method for keratin and mucus; as a counterstain following Heidenhain′s or Weigert′s hematoxylin on paraffin sections; as a substitute for orange G in Flemming′s triple stain.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tarek Alammar et al.
Nanoscale, 5(17), 8045-8055 (2013-07-23)
The phase selective synthesis of nanocrystalline TiO2, titania, in ionic liquids (ILs) is explored. The influence not only of the IL but also of the Ti-precursor, pH, and temperature is investigated. Sonochemical synthesis, microwave synthesis and conventional heating are compared.
Chia-Nan Lin et al.
Environmental science and pollution research international, 19(9), 3743-3750 (2012-10-12)
Solar wastewater treatment based on photocatalytic reactions is a green process that utilizes renewable energy resources and minimizes secondary pollution. Reactor design plays an important role in promoting treatment efficiency and throughput density (based on unit volume of the reactor).
P Kumar et al.
Colloids and surfaces. B, Biointerfaces, 103, 658-661 (2012-12-26)
In this paper, we report on biosynthesis of silver nanoparticles using Ulva lactuca (seaweed) at room temperature along with photocatalytic degradation of methyl orange dye. UV spectral analysis showed peak at 430 nm with special reference to the excitation of
Katarzyna A Michalow et al.
Environmental science and pollution research international, 19(9), 3696-3708 (2012-10-12)
TiO(2):Nb nanopowders within a dopant concentration in the range of 0.1-15 at.% were prepared by one-step flame spray synthesis. Effect of niobium doping on structural, optical and photocatalytic properties of titanium dioxide nanopowders was studied. Morphology and structure were investigated by
Yu Lim Chen et al.
Optics express, 21(6), 7240-7249 (2013-04-03)
A low-cost and efficient photocatalytic reactor for environmental treatment and green technology was presented. ZnO nanorods firmly growing on polycarbonate optical disk substrate are generally perpendicular to the substrate as the immobilized photocatalyst of the spinning disk reactor. The photocatalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service