Skip to Content
Merck
All Photos(1)

Key Documents

291099

Sigma-Aldrich

α-Naphtholbenzein

indicator grade

Synonym(s):

4,4′-(α-Hydroxybenzylidene)di-1-naphthol, p-naphtholbenzein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$158.00
25 G
$539.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$158.00
25 G
$539.00

About This Item

Empirical Formula (Hill Notation):
C27H18O2
CAS Number:
Molecular Weight:
374.43
Beilstein:
2628063
EC Number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$158.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

grade

indicator grade

Quality Level

form

powder

technique(s)

titration: suitable

color

faintly Faint Brown to Very Dark Brown to Very Dark Red and Black

pH

0.0, green
0.8, yellow
10.0, blue-green
8.2, yellow

mp

230-235 °C (lit.)

ε (extinction coefficient)

≥50000 at 207-213 nm in methanol at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(\C(c2ccccc2)=C3\C=CC(=O)c4ccccc34)c5ccccc15

InChI

1S/C27H18O2/c28-25-16-14-23(19-10-4-6-12-21(19)25)27(18-8-2-1-3-9-18)24-15-17-26(29)22-13-7-5-11-20(22)24/h1-17,28H/b27-24-

InChI key

VDDWRTZCUJCDJM-PNHLSOANSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Naphtholbenzein is a dye belonging to the Benzein family insoluble in water and soluble in alcohol. α-Naphtholbenzein is primarily used as an indicator for pH range 8.2-10.0 (mostly in acid-base titrations), causing a color change for the neutral solutions turning alkaline. Its applications include detection of bacterial growth in packed food, viable cells, enzymes, ammonia odors in patients with bacterial infection, and many more.

Application

α-Naphtholbenzein has been used as an indicator during sequential injection analysis technique done for acid-base titrations.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 貴社で販売のSigma-Aldrichのp-ナフトールベンゼインが酸価試験法(ASTM D974, ASTM D3242)のANNEXに記載の仕様を満たしている事を確認させて下さい。 ①外観, ②塩化物, ③溶解性, ④最小吸光度, ⑤pHレンジ

    1 answer
    1. No. Product 291099 does not meet ASTM D974 or ASTM3242. Product 291099 has an M Clarity quality level of 100 - which is the lowest of the 6 quality levels available. The product meets the specifications found on the Specification sheet that is available from the website. The methods have been developed by our company and are not traceable to either ASTM D974 or ASTM3242.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service