Skip to Content
Merck
All Photos(1)

Key Documents

71513

Sigma-Aldrich

Sodium dimethyldithiocarbamate solution

~40% in H2O

Synonym(s):

Dimethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$58.90

$58.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$58.90

About This Item

Linear Formula:
(CH3)2NCSSNa
CAS Number:
Molecular Weight:
143.21
Beilstein:
3569024
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

~40% in H2O

functional group

amine
dithiocarbamate

SMILES string

CN(C)C(=S)S[Na]

InChI

1S/C3H7NS2.Na/c1-4(2)3(5)6;/h1-2H3,(H,5,6);/q;+1/p-1

InChI key

VMSRVIHUFHQIAL-UHFFFAOYSA-M

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service