Skip to Content
Merck
All Photos(1)

Key Documents

21853

Supelco

Carbonate ionophore II solution

Selectophore

Synonym(s):

1-(Dodecylsulfonyl)-4-trifluoroacetylbenzene, ETH 6019

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29F3O3S
CAS Number:
Molecular Weight:
406.50
Beilstein:
4765347
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

SMILES string

CCCCCCCCCCCCS(=O)(=O)c1ccc(cc1)C(=O)C(F)(F)F

InChI

1S/C20H29F3O3S/c1-2-3-4-5-6-7-8-9-10-11-16-27(25,26)18-14-12-17(13-15-18)19(24)20(21,22)23/h12-15H,2-11,16H2,1H3

InChI key

WQOBMOYCYLMGKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

solution in 0.5 mL tetrahydrofuran

Other Notes

Lipophilic, neutral ionophore selective for carboxylate and hydroxide. Used in solvent-polymeric membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Wang
Analytical Chemistry, 63, 970-970 (1991)
C. Behringer
Analytica Chimica Acta, 233, 41-41 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service