Skip to Content
Merck
All Photos(2)

Key Documents

201286

Sigma-Aldrich

Fast Red ITR

Synonym(s):

Azoic Diazo No. 42

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$292.00

$292.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$292.00

About This Item

Empirical Formula (Hill Notation):
C11H18N2O3S
CAS Number:
Molecular Weight:
258.34
Colour Index Number:
37150
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$292.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

>96% (TLC)

Quality Level

form

solid

mp

102-105 °C (lit.)

solubility

chloroform: methanol (1:1): 1%

λmax

226 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCN(CC)S(=O)(=O)c1ccc(OC)c(N)c1

InChI

1S/C11H18N2O3S/c1-4-13(5-2)17(14,15)9-6-7-11(16-3)10(12)8-9/h6-8H,4-5,12H2,1-3H3

InChI key

WBGVVXSCGNGJFL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ana M Martín-Hernández et al.
Journal of virology, 82(8), 4064-4071 (2008-02-15)
RNA silencing is a host defense mechanism that limits the accumulation and spread of viruses in infected plants. Correspondingly, plant viruses encode suppressors of silencing. In the positive-strand RNA virus Tobacco rattle virus (TRV), the suppressor of silencing is a
Florian R Fritzsche et al.
BMC cancer, 8, 381-381 (2008-12-23)
Enhanced activity of histone deacetylases (HDAC) is associated with more aggressive tumour behaviour and tumour progression in various solid tumours. The over-expression of these proteins and their known functions in malignant neoplasms has led to the development of HDAC inhibitors
Jutta Schade et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(2), 147-155 (2007-10-31)
The expression of dipeptidyl peptidase 4 (DP4, CD26) affects T-cell recruitment to lungs in an experimental rat asthma model. Furthermore, the gene of the structural homologous DP10 represents a susceptibility locus for asthma in humans, and the functional homologous DP8/9

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service