Skip to Content
Merck
All Photos(2)

Key Documents

229113

Sigma-Aldrich

Nuclear Fast Red

Synonym(s):

4-Amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$432.00

$432.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$432.00

About This Item

Empirical Formula (Hill Notation):
C14H8NNaO7S
CAS Number:
Molecular Weight:
357.27
Colour Index Number:
60760
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$432.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

ε (extinction coefficient)

≥11000 at 515-540 nm in H2O at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Nc1c(O)c(c(O)c2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H9NO7S.Na/c15-9-7-8(12(18)14(13(9)19)23(20,21)22)11(17)6-4-2-1-3-5(6)10(7)16;/h1-4,18-19H,15H2,(H,20,21,22);/q;+1/p-1

InChI key

IFSXZLJQEKGQAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Nuclear fast red has been used to counterstain nuclei and renal collecting ducts.[1][2]
Suitable for determination of calcium
Used as a biological stain

Biochem/physiol Actions

Nuclear fast red dye has anthraquinonic nature and is used either alone or in combination with other dyes. It is mainly used for staining of nuclei.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Marie Exbrayat
Genome Visualization by Classic Methods in Light Microscopy, Methods in Visualization (2000)
A Mouse Model That Reproduces the Developmental Pathways and Site Specificity of
the Cancers Associated With the Human BRCA1 Mutation Carrier State
Liu Y
EBioMedicine, 2, 1318-1330 (2015)
Preventing Corneal Calcification Associated With Xylazine for Longitudinal Optical Coherence Tomography in Young Rodents.
Zhou TE
Investigative Ophthalmology & Visual Science, 58, 461-469 (2017)
M M Rahman et al.
British journal of cancer, 100(3), 501-510 (2009-01-22)
The molecular pathogenesis of gastroenteropancreatic (GEP) neuroendocrine tumours (NETs) remains to be elucidated. High-mobility group A (HMGA) proteins play important roles in the regulation of transcription, differentiation, and neoplastic transformation. In this study, the expression of HMGA1 and HMGA2 was
Florian R Fritzsche et al.
BMC cancer, 8, 381-381 (2008-12-23)
Enhanced activity of histone deacetylases (HDAC) is associated with more aggressive tumour behaviour and tumour progression in various solid tumours. The over-expression of these proteins and their known functions in malignant neoplasms has led to the development of HDAC inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service