Skip to Content
Merck
All Photos(2)

Key Documents

19448

Supelco

Anhydroerythromycin A

analytical standard

Synonym(s):

Erythromycin anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$361.00

$361.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
2 MG
$361.00

About This Item

Empirical Formula (Hill Notation):
C37H65NO12
CAS Number:
Molecular Weight:
715.91
UNSPSC Code:
41116107
NACRES:
NA.24

$361.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](OC3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)C[C@@H](C)C5(O4)OC1(C)[C@H](O)[C@H]5C

InChI

1S/C37H65NO12/c1-14-25-36(10)29(40)22(6)37(50-36)18(2)16-35(9,49-37)31(48-33-27(39)24(38(11)12)15-19(3)44-33)20(4)28(21(5)32(42)46-25)47-26-17-34(8,43-13)30(41)23(7)45-26/h18-31,33,39-41H,14-17H2,1-13H3/t18-,19-,20+,21-,22-,23+,24+,25-,26+,27-,28+,29-,30+,31-,33+,34-,35-,36-,37+/m1/s1

InChI key

YKAVHPRGGAUFDN-JTQLBUQXSA-N

General description

Anhydroerythromycin A also known as erythromycin anhydride is a macrolide antibiotic, which are basic and lipophilic molecule.[1][2]
Chemical structure: macrolide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Norihide Nakada et al.
Water research, 41(19), 4373-4382 (2007-07-17)
We investigated the efficiencies of removal of 24 pharmaceutically active compounds (PhACs) during sand filtration and ozonation in an operating municipal sewage treatment plant (STP). The target compounds were 2 phenolic antiseptics (thymol, triclosan), 5 acidic analgesics or anti-inflammatories (ibuprofen
Shinwoo Yang et al.
Analytical and bioanalytical chemistry, 385(3), 623-636 (2006-05-23)
Two wastewater treatment plants (WWTPs) of northern Colorado were monitored for anhydroerythromycin and tylosin. An analytical method has been developed and validated for the trace determination and confirmation of these compounds in the raw influent and final effluent water matrices.
Ute I Scholl et al.
The Journal of clinical investigation, 127(7), 2739-2750 (2017-06-13)
Aldosterone-producing adenomas (APAs) are benign tumors of the adrenal gland that constitutively produce the salt-retaining steroid hormone aldosterone and cause millions of cases of severe hypertension worldwide. Either of 2 somatic mutations in the potassium channel KCNJ5 (G151R and L168R

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service