Skip to Content
Merck
All Photos(2)

Key Documents

18566

Supelco

all-cis-4,7,10,13,16-Docosapentaenoic acid

analytical standard

Synonym(s):

(4Z,7Z,10Z,13Z,16Z)-4,7,10,13,16-Docosapentaenoic acid, (all-Z)-4,7,10,13,16-Docosapentaenoic acid, Osbond acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,660.00

$1,660.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,660.00

About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$1,660.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

grade

analytical standard

description

C22:5n6

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O

InChI

1S/C22H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h6-7,9-10,12-13,15-16,18-19H,2-5,8,11,14,17,20-21H2,1H3,(H,23,24)/b7-6-,10-9-,13-12-,16-15-,19-18-

InChI key

AVKOENOBFIYBSA-WMPRHZDHSA-N

General description

Docosapentaenoic acid is a very long-chain polyunsaturated fatty acid, and has two or more double bonds in its acyl chains.[1]

Application

It was used as supplement for fed cells during phospholipid analysis by HPLC/electrospray ionization mass spectrometry (ESI/MS) and HPLC/ESI/MS/MS.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Kuhnt et al.
The Journal of nutrition, 144(4), 447-460 (2014-02-21)
Dietary supplementation with echium oil (EO) containing stearidonic acid (SDA) is a plant-based strategy to improve long-chain (LC) n-3 (ω-3) polyunsaturated fatty acid (PUFA) status in humans. We investigated the effect of EO on LC n-3 PUFA accumulation in blood
J K Beckman et al.
Lipids, 15(6), 389-394 (1980-06-01)
The metabolism of [1-14C] linoleic, [1-14C] arachidonic and [3-14C] docosa-4,7,10,13,16-pentaenoic acids was investigated after intratesticular injection of the labeled compounds and isolation of rat Sertoli and germinal cell. Following injection of either 14C-linoleate or 14C-arachidonate, the specific activity (sp act)
Yuru Deng et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(9), 2866-2871 (2009-05-02)
Very long-chain polyunsaturated fatty acids (VLC-PUFAs), such as docosahexaenoic acid (DHA) and docosapentaenoic acid (DPA), have recently made it to the realm of "magical molecules" based on their multiple presumably beneficial effects in biological systems, making these PUFAs particularly interesting
B S Mohammed et al.
The Biochemical journal, 326 ( Pt 2), 425-430 (1997-09-18)
It is now established that fatty acid 7,10,13,16-22:4 is metabolized into 4,7,10,13,16-22:5 as follows: 7,10,13,16-22:4-->9,12,15, 18-24:4-->6,9,12,15,18-24:5-->4,7,10,13,16-22:5. Neither C24 fatty acid was esterified to 1-acyl-sn-glycero-3-phosphocholine (1-acyl-GPC) by microsomes, whereas the rates of esterification of 4, 7,10,13,16-22:5, 7,10,13,16-22:4 and 5,8,11,14-20:4 were respectively
B S Mohammed et al.
Archives of biochemistry and biophysics, 317(1), 179-184 (1995-02-20)
When rat liver microsomes were incubated with [1-14C]22:4(n-6) under standard conditions for measuring acyl-CoA desaturases, it was not possible to detect the synthesis of any 22:5(n-6). When malonyl-CoA and NADPH were included in the incubation, 22:4(n-6) was chain elongated to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service