Skip to Content
Merck
All Photos(1)

Key Documents

T8626

Sigma-Aldrich

Glyceryl tributyrate

≥99%

Synonym(s):

1,2,3-Tributyrylglycerol, Glycerol tributyrate, Tributyrin

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$95.80
100 ML
$294.00

$95.80

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 10, 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
$95.80
100 ML
$294.00

About This Item

Linear Formula:
(CH3CH2CH2COOCH2)2CHOCOCH2CH2CH3
CAS Number:
Molecular Weight:
302.36
Beilstein:
1714746
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$95.80

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 10, 2025Details


Request a Bulk Order

Assay

≥99%

refractive index

n20/D 1.435 (lit.)

bp

129-131 °C/0.03 mmHg (lit.)
287-288 °C (lit.)

density

1.032 g/mL at 20 °C (lit.)

lipid type

neutral glycerides

SMILES string

CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

InChI

1S/C15H26O6/c1-4-7-13(16)19-10-12(21-15(18)9-6-3)11-20-14(17)8-5-2/h12H,4-11H2,1-3H3

InChI key

UYXTWWCETRIEDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Study on the digestive characteristics of short-and medium-chain fatty acid structural lipid and its rapid intervention on gut microbes: In vivo and in vitro studies.: This paper investigates the digestive properties of glyceryl tributyrate and its effects on gut microbiota, highlighting its significance in nutritional biochemistry (Liu et al., 2022).

  • Molecular Characterization of Novel Family IV and VIII Esterases from a Compost Metagenomic Library.: The characterization of novel esterases, including those acting on glyceryl tributyrate, underscores their potential in environmental and industrial biochemistry (Park et al., 2021).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

345.2 °F - closed cup

Flash Point(C)

174 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelsea R Thorn et al.
Journal of controlled release : official journal of the Controlled Release Society, 319, 168-182 (2019-12-28)
In the advent of the post-antibiotic era, new strategies are urgently required to improve the efficacy of antimicrobials and outsmart multi-drug resistant bacteria. Exploiting a basic survival mechanism of bacteria, lipase production, monoolein liquid crystal nanoparticles (MO-LCNPs) were investigated as
Lonneke Onrust et al.
Veterinary research, 51(1), 56-56 (2020-04-21)
Butyrate has been used extensively as a feed additive to improve gut health and to decrease Salmonella colonization in poultry. Salmonella mainly colonizes the ceca so butyrate concentrations should be increased in this gut segment. Discrepancies on the effects of
M H Sodergren et al.
Scandinavian journal of surgery : SJS : official organ for the Finnish Surgical Society and the Scandinavian Surgical Society, 99(3), 153-161 (2010-11-04)
current studies suggest immunonutrition decreases the inflammatory process, infection rates and reduces length of hospital stay in surgical patients, however studies are often conducted on heterogeneous groups of patients with varying composition of the immuno-nutrition. We aim to investigate the
Peter J Richards et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 92(2), 439-446 (2009-11-21)
Epidural adhesion formation is believed to be a central governing factor in the prevalence of pain after spinal surgery and is regarded as being the primary instigator of neural tethering, leading to complications during revision surgery. In this study, we
Marco Aurélio Ramirez Vinolo et al.
American journal of physiology. Endocrinology and metabolism, 303(2), E272-E282 (2012-05-25)
The aim of this study was to investigate whether treatment with tributyrin (Tb; a butyrate prodrug) results in protection against diet-induced obesity and associated insulin resistance. C57BL/6 male mice fed a standard chow or high-fat diet were treated with Tb

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service