Skip to Content
Merck
All Photos(2)

Documents

17264

Supelco

Methyl cis-11-octadecenoate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
Beilstein:
1911845
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

ester

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCC\C=C/CCCCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-9H,3-7,10-18H2,1-2H3/b9-8-

InChI key

PVVODBCDJBGMJL-HJWRWDBZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cis-11-octadecenoate is a fatty acid methyl ester.

Application

It may be used as standard mixture along with bacterial FAME and polyunsaturated fatty acids to identify the peak during GC analysis to determine fatty acid composition in bacterial cells and lipid extraction.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatty acid profiles, a chemotaxonomic key for the classification of strains of the family Leptospiraceae.
Cacciapuoti, B., L. Ciceroni, and D. Attard Barbini.
International Journal of Systematic and Evolutionary Microbiology, 41.2, 295-300 (1991)
Burkholderia cenocepacia phenotypic clonal variation during a 3.5-year colonization in the lungs of a cystic fibrosis patient.
Coutinho CP
Infection and Immunity, 79(7), 2950-2960 (2011)
D D Bikle et al.
Endocrinology, 114(1), 260-267 (1984-01-01)
To evaluate whether membrane fluidity plays a role in regulating calcium flux across the intestinal brush border, we purified brush border membranes from different regions of the chick intestine and determined the relationship of their ability to transport calcium and
G Karsenty et al.
Pflugers Archiv : European journal of physiology, 403(2), 151-155 (1985-02-01)
In the present work we studied rapid in vitro effects of calcitriol (1,25(OH)2 vitamin D3) on the intestinal transport of inorganic phosphate (Pi). Enterocytes from vitamin D replete (D+) as well as vitamin D depleted (D-) rats were isolated mechanically
R A Harris et al.
Journal of neurochemistry, 44(4), 1274-1281 (1985-04-01)
The effects of membrane perturbants (ethanol, pentobarbital, chloroform, diethylether, phenytoin, cis-vaccenic acid methylester, and cis-vaccenoyl alcohol) on the lipid order of mouse brain synaptic plasma membranes (SPM) were tested by fluorescence polarization using 1,6-diphenyl-1,3,5-hexatriene (DPH) as a probe of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service