Skip to Content
Merck
All Photos(1)

Key Documents

17263

Supelco

Methyl cis-11-eicosenoate

analytical standard

Synonym(s):

cis-11-Eicosenoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$209.00

$209.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$209.00

About This Item

Linear Formula:
CH3(CH2)7HC=CH(CH2)9CO2CH3
CAS Number:
Molecular Weight:
324.54
Beilstein:
1727804
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$209.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

ester

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCC(=O)OC

InChI

1S/C21H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h10-11H,3-9,12-20H2,1-2H3/b11-10-

InChI key

RBKMRGOHCLRTLZ-KHPPLWFESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl cis-11-eicosenoate is a fatty acid methyl ester.[1]

Application

It may be used as FAME standard to locate double bonds in polyunsaturated fatty acid methyl esters using ion-trap mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in phospholipase A2 activity and lipid content during early development of Atlantic halibut (Hippoglossus hippoglossus).
Evans, R. P., et al.
Marine biology, 130.3, 369-376 (1998)
Acetonitrile chemical ionization tandem mass spectrometry to locate double bonds in polyunsaturated fatty acid methyl esters.
Van Pelt CK, Brenna JT.
Analytical Chemistry, 71(10), 1981-1989 (1999)
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service