Skip to Content
Merck
All Photos(1)

Documents

17134

Millipore

CPC-Agar (Base)

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Cellobiose Polymyxin Colistin Agar (Base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

composition

agar, 15 g/L
beef extract, 5 g/L
bromothymol blue, 0.04 g/L
cellobiose, 15 g/L
cresol red, 0.04 g/L
peptic digest of animal tissue, 10 g/L
sodium chloride, 20 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.6±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

selective and differential for Vibrio spp.

General description

Cellobiose-polymyxin B-colistin(CPC) agar is a medium used for the isolation and differentiation of Vibrio vulnificus in laboratory testing.

Application

for the cultivation and identification of Vibrio species from foods

Preparation Note

Suspend 32.54 g in 500 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 45°C and aseptically add reconstituted contents of 1 vial of CPC Supplement (Cat. No. 17137). Mix well.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved isolation of Vibrio vulnificusfrom seawater and sediment with cellobiose-colistin agar.
H?i L, et al.
Applied and Environmental Microbiology, 64(5), 1721-1724 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service