The proprietary chromogenic mixture incorporated in the medium helps in the chromogenic differentiation of Vibrio cholerae and Vibrio parahaemolyticus. The high (alkaline) pH of the medium helps in selective isolation of Vibrio species.
92323
Vibrio ChromoSelect Agar
NutriSelect® Plus, suitable for microbiology
Select a Size
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 15.0 g/L
chromogenic mixture, 5.5 g/L
peptic digest of animal tissue, 10.0 g/L
sodium chloride, 25.0 g/L
sodium cholate, 1.0 g/L
sodium citrate, 6.0 g/L
sodium thiosulfate, 5.0 g/L
manufacturer/tradename
NutriSelect® Plus
final pH
8.5±0.2 (25 °C)
application(s)
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
selective and differential for Vibrio spp.
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.
-
What Vibrio species is the HiCrome TM Vibrio Agar selective for?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service