Skip to Content
Merck
All Photos(1)

Key Documents

14509

Sigma-Aldrich

Poly(ethylene glycol) bis(amine)

Mw 20,000, carboxyl reactive, amine

Synonym(s):

Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol) bis(amine), Mw 20,000

mol wt

Mw 20,000

Quality Level

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

With HCl, reduction agent; stereoselective allylation of carbonyl compounds; in situ generation of tinenolates for directed aldol reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yamamoto, Y.; Asao, N.
Chemical Reviews, 93, 2207-2207 (1993)
Huahua Jian et al.
The Journal of organic chemistry, 68(13), 5091-5103 (2003-06-21)
Four caltrop-shaped molecules that might be useful as surface-bound electric field-driven molecular motors have been synthesized. The caltrops are comprised of a pair of electron donor-acceptor arms and a tripod base. The molecular arms are based on a carbazole or
Gaëlle-Anne Cremer et al.
Journal of peptide science : an official publication of the European Peptide Society, 12(6), 437-442 (2006-01-25)
This paper describes the optimization of a synthesis of a difficult sequence related to a 12-mer sequence of a Pan DR epitope (PADRE). Elongation was followed by on-line monitoring of the N(alpha)-Fmoc removal adapted for the batch methodology. Studying the
Stephen J Connon et al.
Bioorganic & medicinal chemistry letters, 12(14), 1873-1876 (2002-06-28)
The synthesis and olefin metathesis activity in protic solvents of 7, a phosphine-free ruthenium alkylidene bound to a hydrophilic solid support are reported. This heterogeneous catalyst promotes relatively efficient ring closing- and cross-metathesis reactions in both methanol and water. The
C S Lee et al.
Artificial organs, 21(9), 1002-1006 (1997-09-01)
Various modifications of alginate-poly-L-lysine microcapsules were made, such as the inclusion of polyethylenimine (PEI) or carboxyl methyl cellulose (CMC) in the core and the coating of bis(polyoxyethylene bis[amine]) (PEGA) onto the microcapsule membrane surface. A characterization of the modified microcapsules

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service