Skip to Content
Merck
All Photos(1)

Key Documents

12600

Sigma-Aldrich

Potassium monopersulfate triple salt

≥47% KHSO5 basis

Synonym(s):

Oxone®, Potassium peroxymonosulfate, ‘Caro’s acid’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2KHSO5 · KHSO4 · K2SO4
CAS Number:
Molecular Weight:
307.38
MDL number:
UNSPSC Code:
12352120
PubChem Substance ID:

form

solid

reaction suitability

reagent type: oxidant

concentration

≥4% (active oxygen)
≥47% (KHSO5)

SMILES string

[K+].[K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.OS(=O)(=O)O[O-].OS(=O)(=O)O[O-]

InChI

1S/5K.2H2O5S.2H2O4S/c;;;;;2*1-5-6(2,3)4;2*1-5(2,3)4/h;;;;;2*1H,(H,2,3,4);2*(H2,1,2,3,4)/q5*+1;;;;/p-5

InChI key

HJKYXKSLRZKNSI-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
  • Oxidative umpolung of alkali metal bromides
  • Controlled epoxidation reactons
  • Desulfurization reactions for fluorescent chemosignaling (fluorescent turn-on type signaling)

Usees:
  • Oxidizing agent (oxidant)
  • Terminal oxidant for for bromolactonization of alkenoic acids

Legal Information

OXONE is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hsiao-Ying Chou et al.
Polymers, 12(9) (2020-09-24)
Polymer-based hydrogels demonstrate superior performance when used as wound dressing. An ideal dressing should possess an active healing function, absorb wound exudates, and provide a moist interface on the wound for rapid injury repair and the prevention of pain and
Bana Shriky et al.
Journal of colloid and interface science, 565, 119-130 (2020-01-17)
Understanding structure-property relationships is critical for the development of new drug delivery systems. This study investigates the properties of Pluronic smart hydrogel formulations for future use as injectable controlled drug carriers. The smart hydrogels promise to enhance patient compliance, decrease
Elisamaria Tasca et al.
Journal of colloid and interface science, 579, 551-561 (2020-07-06)
Doxorubicin hydrochloride (DX) is widely used as a chemotherapeutic agent, though its severe side-effects limit its clinical use. A way to overcome these limitations is to increase DX latency through encapsulation in suitable carriers. However, DX has a high solubility
Masami Fukushima et al.
Chemosphere, 80(8), 860-865 (2010-06-19)
Tetrabromobisphenol A (TBBPA), a commercially used brominated flame retardant, also functions as an endocrine disruptor and is of serious concern in terms of environmental pollution. TBBPA has been detected in leachates from landfills, because hydrophobic interactions with humic acids (HAs)
Raf Dewil et al.
Journal of hazardous materials, 146(3), 577-581 (2007-05-29)
During the anaerobic digestion of wastewater treatment sludge, commonly called biosolids, an energy rich biogas is formed which is now considered as renewable energy source and widely used for the production of heat and/or electricity. Pre-treatment methods, which achieve a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service