Skip to Content
Merck
All Photos(3)

Key Documents

105902

Sigma-Aldrich

p-Toluenesulfonamide

reagent grade, 97%

Synonym(s):

4-Methylbenzene-1-sulfonamide, p-Tosylamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$58.30
1 KG
$113.00

$58.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$58.30
1 KG
$113.00

About This Item

Linear Formula:
CH3C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22
Beilstein:
472689
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Quality Level

Assay

97%

form

solid

mp

134-137 °C (lit.)

solubility

DMSO: soluble

functional group

sulfonamide

SMILES string

Cc1ccc(cc1)S(N)(=O)=O

InChI

1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)

InChI key

LMYRWZFENFIFIT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

p-Toluenesulfonamide undergoes FeCl3-catalyzed direct substitution reaction with benzylic and allylic alcohols[1].It is employed as nucleophile in tetrabutylammonium fluoride (TBAF) catalyzed vinyl aziridine opening reaction[1].

Application

p-Toluenesulfonamide was used to prepare the precursor required for synthesis of ethyl 6-phenyl-1-tosyl-1,2,5,6-tetrahydropyridine-3-carboxylate[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

395.6 °F - closed cup

Flash Point(C)

202 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHOSPHINE-CATALYZED [4+2] ANNULATION: SYNTHESIS OF ETHYL 6-PHENYL-1-TOSYL-1,2,5,6-TETRAHYDROPYRIDINE-3-CARBOXYLATE.
Kui Lu et al.
Organic syntheses; an annual publication of satisfactory methods for the preparation of organic chemicals, 2009(86), 212-224 (2010-02-18)
An efficient FeCl3 -catalyzed amidation reaction of secondary benzylic and allylic alcohols with carboxamides or p-toluenesulfonamide.
Jana U, et al.
Tetrahedron Letters, 42(37), 6433-6435 (2001)
Charles H Reynolds et al.
Journal of medicinal chemistry, 51(8), 2432-2438 (2008-04-03)
Ligand efficiency (i.e., potency/size) has emerged as an important metric in drug discovery. In general, smaller, more efficient ligands are believed to have improved prospects for good drug properties (e.g., bioavailability). Our analysis of thousands of ligands across a variety
Yang Yang et al.
Organic letters, 13(20), 5608-5611 (2011-09-16)
BF(3)·OEt(2)-catalyzed direct cyanation of indoles and pyrroles using a less toxic, bench-stable, and easily handled electrophilic cyanating agent N-cyano-N-phenyl-para-toluenesulfonamide (NCTS) affords 3-cyanoindoles and 2-cyanopyrroles in good yields with excellent regioselectivity. The substrate scope is broad with respect to indoles and
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 15(4), 971-976 (2005-02-03)
The inhibition of a newly cloned human carbonic anhydrase (CA, EC 4.2.1.1), isozyme VII (hCA VII), has been investigated with a series of aromatic and heterocyclic sulfonamides, including some of the clinically used derivatives (acetazolamide, methazolamide, ethoxzolamide, dichlorophenamide, dorzolamide, brinzolamide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service