Skip to Content
Merck
All Photos(2)

Key Documents

124508

Sigma-Aldrich

Gallocyanine

Dye content 90 %

Synonym(s):

7-Dimethylamino-4-hydroxy-3-oxo-phenoxazine-1-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$194.00

$194.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$194.00

About This Item

Empirical Formula (Hill Notation):
C15H13ClN2O5
CAS Number:
Molecular Weight:
336.73
Colour Index Number:
51030
Beilstein:
307454
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$194.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

form

powder

Quality Level

composition

Dye content, 90%

solubility

1 N NH4OH: 10 mg/mL

λmax

601 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].C\[N+](C)=C1\C=CC2=Nc3c(OC2=C1)c(O)c(O)cc3C(O)=O

InChI

1S/C15H12N2O5.ClH/c1-17(2)7-3-4-9-11(5-7)22-14-12(16-9)8(15(20)21)6-10(18)13(14)19;/h3-6H,1-2H3,(H2-,16,18,19,20,21);1H

InChI key

ADAUKUOAOMLVSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gallocyanine belongs to the oxazine class of dyes. It is synthesised from methanol solution of gallic acid in the presence of excess iodine.[1] Gallocyanine is used to stain animal cells,[2] mostly the nucleic acids.[3] It also acts as a chelating agent and is used to determine bioactive compounds and several metal ions.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christa Gaskill et al.
Pulmonary circulation, 6(4), 483-497 (2017-01-17)
Rapid access to lung-derived cells from stable subjects is a major challenge in the pulmonary hypertension field, given the relative contraindication of lung biopsy. In these studies, we sought to demonstrate the importance of evaluating a cell type that actively
V V Zadorozhnyĭ
Arkhiv patologii, 44(11), 67-68 (1982-01-01)
It is suggested that a nuclear stain; gallocyanine, be used simultaneously with the three main histological staining methods: hematoxylin-eosin, Van-Gieson's, and Nissl' stainings. This modification is technically simple, gives preparations of a high quality, and is time- and labour-saving.
Lawei Yang et al.
Cell death & disease, 11(10), 827-827 (2020-10-06)
Idiopathic pulmonary fibrosis (IPF) is a progressive interstitial lung disease characterized by chronic non-specific inflammation of the interstitial lung and extensive deposition of collagen fibers leading to destruction of lung function. Studies have demonstrated that exposure to fine particulate matter
P Damen et al.
Mechanisms of development, 56(1-2), 41-60 (1996-05-01)
During early development of the gastropod mollusc Patella vulgata, the stem cell of the mesoderm (3D-macromere) is induced. As a result of this induction, the embryo becomes dorsoventrally organised. At about the same time in development, ciliated cells, so-called trochoblasts
C-banding-like pattern of mouse Feulgen and gallocyanin chrome alum stained chromosomes.
S Garagna et al.
Basic and applied histochemistry, 26(4), 303-304 (1982-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service