Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Gallocyanine
Synonym(s):
7-Dimethylamino-4-hydroxy-3-oxo-phenoxazine-1-carboxylic acid
Empirical Formula (Hill Notation):
C
15
H
13
ClN
2
O
5
CAS Number:
1562-85-2
Molecular Weight:
336.73
EC Number:
216-345-7
Products
Product Category
stains
Brand
Sigma-Aldrich
Physical Form
Quality Segment
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
124508
Gallocyanine,
Dye content 90 %
Expand
View Pricing
An unknown error has occured.