Skip to Content
Merck
All Photos(1)

Key Documents

11491

Supelco

Cobalt(II) ionophore IV

Selectophore, function tested

Synonym(s):

4-tert-Butylthiacalix[4]arene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H48O4S4
CAS Number:
Molecular Weight:
721.07
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

quality

function tested

SMILES string

CC(C)(C)c1cc2sc3cc(cc(sc4cc(cc(sc5cc(cc(sc(c1)c2O)c5O)C(C)(C)C)c4O)C(C)(C)C)c3O)C(C)(C)C

InChI

1S/C40H48O4S4/c1-37(2,3)21-13-25-33(41)26(14-21)46-28-16-23(39(7,8)9)18-30(35(28)43)48-32-20-24(40(10,11)12)19-31(36(32)44)47-29-17-22(38(4,5)6)15-27(45-25)34(29)42/h13-20,41-44H,1-12H3

InChI key

PDEJSTNRUYUEQL-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electroanalytical studies on cobalt(II) selective potentiometric sensor based on bridge modified calixarene in poly(vinyl chloride)
Gupta, V.K., et al.
Electrochimica Acta, 53, 5409-5414 (2008)
Zong-Jie Guan et al.
Chemical science, 10(11), 3360-3365 (2019-04-19)
We report the synthesis and structures of two 34-atom metal nanoclusters, namely [Ag34(BTCA)3(C[triple bond, length as m-dash]CBu

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service