Skip to Content
Merck
All Photos(1)

Documents

110078

Sigma-Aldrich

Acetaldehyde

ReagentPlus®, 99%

Synonym(s):

Ethanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
Beilstein:
505984
EC Number:
MDL number:
UNSPSC Code:
12352114
eCl@ss:
39021102
PubChem Substance ID:

biological source

synthetic

Quality Level

vapor density

1.52 (vs air)

vapor pressure

14.63 psi ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

365 °F

expl. lim.

60 %

refractive index

n20/D 1.332 (lit.)

bp

21 °C (lit.)

mp

−125 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-38.0 °F - closed cup

Flash Point(C)

-38.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuji Yokoyama et al.
Alcoholism, clinical and experimental research, 29(4), 622-630 (2005-04-19)
Elevated mean corpuscular volume (MCV) is a traditional biological marker for alcohol abuse and alcoholism, but the underlying mechanism is unclear. Three recent epidemiologic studies consistently showed that MCV was elevated by alcohol drinking more markedly among individuals with genetically
Mikko Salaspuro
Journal of digestive diseases, 12(2), 51-59 (2011-03-16)
Aldehyde dehydrogenase (ALDH2) and alcohol dehydrogenase (ADH) gene polymorphisms associating with enhanced acetaldehyde exposure and markedly increased cancer risk in alcohol drinkers provide undisputable evidence for acetaldehyde being a local carcinogen not only in esophageal but also in gastric cancer.
Marjie L Hard et al.
Placenta, 24(2-3), 149-154 (2003-02-05)
Significant interindividual variability exists following maternal alcohol consumption; not all children born to alcoholic women manifest the symptoms associated with foetal alcohol spectrum disorder (FASD). To investigate the potential role of the placenta as a source of variability by determining
Neil R McGregor
Alcohol (Fayetteville, N.Y.), 41(7), 469-478 (2007-11-06)
Recent introduction of several commercial Kudzu root (Pueraria lobata) containing hangover remedies has occurred in western countries. The available data is reviewed to assess if there are any potential concerns in relationship to the development of neoplasm if these products
Keitaro Matsuo et al.
Carcinogenesis, 34(7), 1510-1515 (2013-03-05)
The impact of alcohol on the risk of stomach cancer is controversial. Although aldehyde dehydrogenase 2 (ALDH2) Glu504Lys (rs671) polymorphism has a strong effect on acetaldehyde metabolism, little is known about its impact on stomach cancer risk when combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service