Skip to Content
Merck
All Photos(1)

Documents

8.00004

Sigma-Aldrich

Acetaldehyde

for synthesis

Synonym(s):

Acetaldehyde, Ethanal, Acetic Aldehyde, Ethyl Aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12352114
EC Index Number:
200-836-8
NACRES:
NA.22

vapor pressure

1006 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

140 °C

expl. lim.

4-57 % (v/v)

pH

5 (20 °C, 10 g/L in H2O)

bp

20.4 °C/1013 hPa

mp

-123 °C

transition temp

flash point -20 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

General description

Acetaldehyde is an organic compound and one of the significant aldehydes. Acetaldehyde is employed to produce acetic anhydride, acetic acid, cellulose acetate, pyridine derivatives, pentaerythritol, peracetic acid, and terephthalic acid. Furthermore, it is used as a solvent.

Application

Acetaldehyde is used as a raw material to synthesis acetal(1,1-dimetoxyethane) using Smopex 101 fibres catalyst.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 5°C/ 4°C): 0.798 - 0.801

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-38.0 °F - closed cup

Flash Point(C)

-38.89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetaldehyde dimethylacetal synthesis with Smopex 101 fibres as catalyst/adsorbent
Gandi GK, et al.
Chemical Engineering Science, 62(4), 907-918 (2007)
Acetaldehyde
Hagemeyer, HJ
Kirk-Othmer Encyclopedia of Chemical Technology (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service