Skip to Content
Merck
All Photos(1)

Key Documents

09236

Supelco

β-Amyrin

analytical standard

Synonym(s):

β-Amyrenol, Olean-12-en-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,040.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,040.00

About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Beilstein:
2063468
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]12CC(C)(C)CC[C@]1(C)CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C

InChI

1S/C30H50O/c1-25(2)15-16-27(5)17-18-29(7)20(21(27)19-25)9-10-23-28(6)13-12-24(31)26(3,4)22(28)11-14-30(23,29)8/h9,21-24,31H,10-19H2,1-8H3/t21-,22-,23+,24-,27+,28-,29+,30+/m0/s1

InChI key

JFSHUTJDVKUMTJ-QHPUVITPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Amyrin belongs to the group of naturally occurring pentacyclic triterpenes, known for its anti-inflammatory, anti-microbial, anti-oxidative, and chemoprotective properties.

Application

This analytical standard can also be used as follows:

  • Development and validation of two high-performance thin layer chromatography (HPTLC) methods to quantify four biomarkers— β-amyrin, β-sitosterol, lupeol, and ursolic acid in the ethanol extract of G. senegalensis leaves
  • Analysis of methanol extracts of leaf samples collected from five Ficus species by a novel HPTLC densitometric method, for the determination of β-amyrin, validated according to the International Conference on Harmonization (ICH) guidelines
  • Quantification of β-amyrin in the ethanolic extracts collected from two Maytenus species, M. obscura and M. parviflora by a validated HPTLC-densitometric method
  • Secondary metabolite profiling of various plant parts collected from 82 plants belonging to 21 different cannabis strains using gas chromatography-mass spectrometry (GC-MS) for sterols and terpenoids (mono-, sesqui-, tri-), and high-performance liquid chromatography (HPLC) with UV and mass spectrometric (MS) detection for flavonoids

Biochem/physiol Actions

Triterpene that has broad-spectrum analgesic properties for which the underlying mechanism is poorly understood.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Miao Wu et al.
Genes, 11(2) (2020-02-14)
Polyploid plants are more often invasive species than their diploid counterparts. As the invasiveness of a species is often linked to its production of allelopathic compounds, we hypothesize that differences in invasive ability between cytotypes may be due to their
Separation and identification of some common isomeric plant triterpenoids by thin-layer chromatography and high-performance liquid chromatography.
Martelanc M, et al.
Journal of Chromatography A, 1216(38), 6662-6670 (2009)
β-Amyrin induces angiogenesis in vascular endothelial cells through the Akt/endothelial nitric oxide synthase signaling pathway.
Ishii M, et al.
Biochemical and Biophysical Research Communications, 467(4), 676-682 (2015)
Cyril Brendolise et al.
The FEBS journal, 278(14), 2485-2499 (2011-05-18)
The pentacyclic triterpenes, in particular ursolic acid and oleanolic acid and their derivatives, exist abundantly in the plant kingdom, where they are well known for their anti-inflammatory, antitumour and antimicrobial properties. α-Amyrin and β-amyrin are the precursors of ursolic and
Jianhong Ching et al.
Journal of separation science, 35(1), 53-59 (2011-12-01)
Conventional methods of drug discovery from natural products include bioassay-guided fractionation, which is tedious and has low efficiency. The aim of this work is to develop a platform method to rapidly identify bioactive compounds from crude plant extracts and their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service