Skip to Content
Merck
All Photos(1)

Key Documents

SML2232

Sigma-Aldrich

α-Spinasterol

≥98% (HPLC)

Synonym(s):

α-Spinasterin, (3β,5α,22E)-Stigmasta-7,22-dien-3-ol, (E)-5α-Stigmasta-7,22-dien-3β-ol, 5α-Stigmasta-7,22-dien-3β-ol, Bessisterol, Hitodesterol, Spinasterol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$596.00

$150.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$596.00

About This Item

Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

biological source

Aster scaber

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

ethanol: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@H](C)/C=C/[C@@H](CC)C(C)C)([H])C3=CC[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC1

InChI

1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-9,11,19-23,25-27,30H,7,10,12-18H2,1-6H3/b9-8+/t20-,21-,22+,23+,25-,26+,27+,28+,29-/m1/s1

InChI key

JZVFJDZBLUFKCA-FXIAWGAOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Spinasterol is a blood-brain barrier-permeable and orally active plant phytosterol with antioxidant, anticonvulsant (0.1-1 mg/kg i.p. in mice), antidepressant (1-2 mg/kg i.p. in mice) and anti-inflammatory (0.001-1 mg/kg, i.p. or 1-10 mg/kg, i.g. in mice) properties, as well as antinociceptive efficacy via dual actions against transient receptor potential vanilloid 1 (TrpV1; IC50/Emax = 1.4 μM/67% against 2 nM RTX for binding mouse spinal cord membranes; IC50/Emax = 40 μM/62% against 20 μM capsaicin-induced mouse spinal cord synaptosomes Ca2+ influx) and cyclooxygenases (IC50 = 16.17 μM/COX1 and 7.76μM/COX2). a-spinasterol can directly affect membrane structure and packing in a manner similar to cholesterol, both a-spinasterol and stitosterol are reported to lower plasma/liver cholesterol levels and increase fecal cholesterol excretion when supplemented in daily diet (1%) in mice in vivo.
α-Spinasterol is a constituent of argan oil. It regulates nuclear receptors expression and modulates mitochondrial function. It mimics natural cholesterol and its functionality.[1] α-Spinastero lelicits cytoprotective and antiulcerogenic functionality.[2] It has therapeutic potential towards diabetic nephropathy.[3]
Orally active phytosterol with anticonvulsant, antidepressant, anti-inflammatory, and antinociceptive efficacy via dual actions against TrpV1 and COX1/2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the antidepressant-and anxiolytic-like activity of alpha-spinasterol, a plant derivative with TRPV1 antagonistic effects, in mice
Socala K and Wlaz P
Behavioural Brain Research, 303, 19-25 (2016)
The Potential of alpha-Spinasterol to Mimic the Membrane Properties of Natural Cholesterol
Haralampiev I, et al.
Molecules (Basel), 22(8), 1390-1390 (2017)
alpha-Spinasterol isolated from the root of Phytolacca americana and its pharmacological property on diabetic nephropathy
Jeong SI, et al.
Planta Medica, 70(08), 736-739 (2004)
Katarzyna Socała et al.
Journal of neural transmission (Vienna, Austria : 1996), 122(9), 1239-1247 (2015-03-13)
α-Spinasterol is a plant-derived compound which was reported to act as a selective antagonist for the transient receptor potential vanilloid 1 (TRPV1) receptor. Several studies revealed that the TRPV1 receptors might modulate seizure activity in animal models of seizures and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service