Skip to Content
Merck
All Photos(2)

Documents

07562

Supelco

17β(H),21β(H)-Hopane solution

0.1 mg/mL in isooctane, analytical standard

Synonym(s):

A′-Neogammacerane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52
CAS Number:
Molecular Weight:
412.73
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

0.1 mg/mL in isooctane

application(s)

food and beverages

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@]3(C)[C@]([H])(CC[C@]4([H])[C@@]5(C)CCCC(C)(C)[C@]5([H])CC[C@@]34C)[C@@]1(C)CC[C@@H]2C(C)C

InChI

1S/C30H52/c1-20(2)21-12-17-27(5)22(21)13-18-29(7)24(27)10-11-25-28(6)16-9-15-26(3,4)23(28)14-19-30(25,29)8/h20-25H,9-19H2,1-8H3/t21-,22+,23+,24-,25-,27+,28+,29-,30-/m1/s1

InChI key

ZRLNBWWGLOPJIC-PYQRSULMSA-N

General description

17β(H),21β(H)-Hopanes belong to the hopanes family of hydrocarbons which are known as the biomarkers of petroleum products.

Application

17β(H),21β(H)-Hopane may be used as an internal injection standard (IIS) for the determination of triterpanes in riverine, estuarine, and marine sediments in Thailand and in stranded tar-balls on the coasts of Peninsular Malaysia by gas chromatography-quadrupole mass spectrometry (GC-MS).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Irei et al.
Journal of analytical methods in chemistry, 2017, 3504274-3504274 (2017-10-28)
To study the origins of airborne particulate organic matter in southern Ontario, molecular marker concentrations were studied at Hamilton, Simcoe, and York Gateway Tunnel, representing industrial, rural, and heavy traffic sites, respectively. Airborne particulate matter smaller than 10
Sources of polycyclic aromatic hydrocarbons (PAHs) in street dust in a tropical Asian mega-city, Bangkok, Thailand.
Boonyatumanond R, et al.
The Science of the Total Environment, 384(1-3), 420-432 (2007)
Polycyclic aromatic hydrocarbon (PAHs) and hopanes in stranded tar-balls on the coasts of Peninsular Malaysia: applications of biomarkers for identifying sources of oil pollution.
Zakaria MP, et al.
Marine Pollution Bulletin, 42(12), 1357-1366 (2001)
Hind Sbihi et al.
Environmental health : a global access science source, 12, 48-48 (2013-06-21)
Exposure to traffic-related air pollution (TRAP) can adversely impact health but epidemiologic studies are limited in their abilities to assess long-term exposures and incorporate variability in indoor pollutant infiltration. In order to examine settled house dust levels of hopanes, engine
Distribution and origins of polycyclic aromatic hydrocarbons (PAHs) in riverine, estuarine, and marine sediments in Thailand.
Boonyatumanond R, et al.
Marine Pollution Bulletin, 52(8), 942-956 (2006)

Articles

We provide high-quality, relevant hopane reference standards to be used as biomarkers for petrochemical oil field remediation and spill analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service