Skip to Content
Merck
All Photos(2)

Key Documents

06773

Supelco

Caffeic acid

matrix substance for MALDI-MS, ≥99.0% (HPLC), powder, light beige

Synonym(s):

3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
1954563
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

form

powder

color

light beige

mp

211-213 °C (dec.) (lit.)

cation traces

Ba: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for UV test
suitable for matrix substance for MALDI-MS

SMILES string

OC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+

InChI key

QAIPRVGONGVQAS-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A natural dietary phenolic compound found in plants that is an anti-oxidant. Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.
Anti-oxidant phenolic compound found in plants; Inhibits the synthesis of leukotrienes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ren-Wang Jiang et al.
Current medicinal chemistry, 12(2), 237-246 (2005-01-11)
Caffeic acid (3,4-dihydroxycinnamic acid), one of the most common phenolic acids, frequently occurs in fruits, grains and dietary supplements for human consumption as simple esters with quinic acid or saccharides, and are also found in traditional Chinese herbs. Caffeic acid
Jun Liu et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 51, 310-316 (2012-10-27)
The antioxidant activities of ethanolic extract from edible mushroom Agaricus bisporus (A. bisporus) were evaluated by various methods in vitro and in vivo. In antioxidant assays in vitro, ethanolic extract of A. bisporus was found to have strong reducing power
Emmanoel Vilaça Costa et al.
Journal of natural products, 73(6), 1180-1183 (2010-05-19)
Phytochemical investigation of the bark of Guatteria hispida afforded three new alkaloids, 9-methoxy-O-methylmoschatoline (1), 9-methoxyisomoschatoline (2), and isocerasonine (3), along with 10 known alkaloids, 8-oxopseudopalmatine (4), O-methylmoschatoline (5), lysicamine (6), liriodenine (7), 10-methoxyliriodenine (8), nornuciferine (9), anonaine (10), xylopine (11)
Susanne Vogel et al.
Journal of natural products, 71(7), 1237-1241 (2008-07-10)
The minor hop ( Humulus lupulus) chalcones 3'-geranylchalconaringenin (3), 5'-prenylxanthohumol (4), flavokawin (5), xanthohumol H (8), xanthohumol C (9), and 1'',2''-dihydroxanthohumol C (10) were synthesized. The non-natural chalcones 3'-geranyl-6'-O-methylchalconaringenin (2), 3'-methylflavokawin (6), and 2'-O-methyl-3'-prenylchalconaringenin (7) were also synthesized. Cytotoxicity was
Morshed A Chowdhury et al.
Journal of medicinal chemistry, 52(6), 1525-1529 (2009-03-20)
A novel class of 1-(4-methanesulfonylphenyl and 4-aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole hybrid cyclooxygenase-2 (COX-2)/5-lipoxygenase (5-LOX) inhibitory anti-inflammatory agents was designed. Replacement of the tolyl ring present in celecoxib by the N-difluoromethyl-1,2-dihydropyrid-2-one moiety provided compounds showing dual selective COX-2/5-LOX inhibitory activities. 1-(4-Aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole exhibited good anti-inflammatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service