Merck
  • Alkaloids from the bark of Guatteria hispida and their evaluation as antioxidant and antimicrobial agents.
Alkaloids from the bark of Guatteria hispida and their evaluation as antioxidant and antimicrobial agents.
Journal of natural products (2010-05-19)
Emmanoel Vilaça Costa, Maria Lúcia B Pinheiro, Andersson Barison, Francinete R Campos, Marcos J Salvador, Beatriz Helena L N S Maia, Elaine C Cabral, Marcos N Eberlin
ABSTRACT
Phytochemical investigation of the bark of Guatteria hispida afforded three new alkaloids, 9-methoxy-O-methylmoschatoline (1), 9-methoxyisomoschatoline (2), and isocerasonine (3), along with 10 known alkaloids, 8-oxopseudopalmatine (4), O-methylmoschatoline (5), lysicamine (6), liriodenine (7), 10-methoxyliriodenine (8), nornuciferine (9), anonaine (10), xylopine (11), coreximine (12), and isocoreximine (13). The major compounds, 2, 6, 12, and 13, showed significant antioxidant capacity in the ORAC(FL) assay. Compounds 5, 6, and 7 were active against S. epidermidis and C. dubliniensis, with MIC values in the range 12.5-100 microg mL(-1).
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Pricing and availability is not currently available.
Isoquercitrin, primary reference standard
Pricing and availability is not currently available.
Supelco
Quercetin 3-glucoside, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Pricing and availability is not currently available.