Skip to Content
Merck
All Photos(1)

Key Documents

05138

Millipore

Vegetable Extract

suitable for microbiology

Synonym(s):

Meat extract replacement

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$140.00

$140.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$140.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.85

$140.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3%
total nitrogen (N), ≥9%

impurities

≤15% ash

loss

≤7.0% loss on drying

pH

7.0±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, light yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

General description

Vegetable extract is a meat-free alternative to traditional peptones. With a wide distribution of peptides it meets the requirements for replacing Peptone from meat, enzymatic digest (Cat. No. 82962) to yield maximum growth of fastidious microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service