Skip to Content
Merck
All Photos(2)

Documents

02536

Supelco

Fluoride ionophore I

Selectophore

Synonym(s):

Bis(fluorodioctylstannyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H70F2Sn2
CAS Number:
Molecular Weight:
742.33
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

SMILES string

CCCCCCCC[Sn](F)(CCCCCCCC)C[Sn](F)(CCCCCCCC)CCCCCCCC

InChI

1S/4C8H17.CH2.2FH.2Sn/c4*1-3-5-7-8-6-4-2;;;;;/h4*1,3-8H2,2H3;1H2;2*1H;;/q;;;;;;;2*+1/p-2

InChI key

KNHZPNMTFJBTPD-UHFFFAOYSA-L

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Other Notes

Selective ionophore used for the determination of fluoride

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Chaniotakis, K. Jurkschat et al.
Inorganic Chemistry, 2283-2283 (2004)
K. Perdikaki, K. Jurkschat et al.
Analytica Chimica Acta, 467, 197-197 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service