Skip to Content
Merck
All Photos(1)

Key Documents

02138

Supelco

Bis(2-ethylhexyl) adipate

Selectophore, ≥99.0%

Synonym(s):

Adipic acid di(2-ethylhexyl) ester, DOA

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$38.10
5 ML
$104.00
25 ML
$374.00

$38.10


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$38.10
5 ML
$104.00
25 ML
$374.00

About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
Beilstein:
1803774
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$38.10


Estimated to ship onMay 20, 2025


Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (GC)
≥99.0%

refractive index

n20/D 1.447 (lit.)

bp

175 °C/2 mmHg (lit.)

mp

<−70 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer employed in polymeric membrane electrodes[1][2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Ghauri et al.
The Analyst, 119(11), 2323-2326 (1994-11-01)
A macrotetrolide nonactin-based ammonium ionophore (AI) preparation containing 25% nonactin optimized for use in liquid membrane ion-selective electrodes was investigated for its possible use in poly(vinyl chloride) (PVC) matrix membrane-type electrodes. Nine different plasticizing solvent mediators were employed in the
R. Eugster et al.
Analytica Chimica Acta, 289, 1-1 (1994)
Kirsten Jarfelt et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 505-515 (2005-03-08)
Di(2-ethylhexyl) phthalate (DEHP) is a well-known testicular toxicant inducing adverse effects in androgen responsive tissues. Therefore, di(2-ethylhexyl) adipate (DEHA) is currently being evaluated as a potential substitute for DEHP. Similarities in structure and metabolism of DEHP and DEHA have led
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 517-525 (2005-03-08)
This study aimed to characterize the effects of di(2-ethylhexyl) phthalate (DEHP) on the fetal rat testes and relate them to the effects seen in adults. Histopathological effects in fetal testes were examined with immunohistochemistry for anti-Mullerian hormone (AMH), 3beta-hydroxysteroid dehydrogenase
Yadollah Yamini et al.
Journal of separation science, 32(18), 3201-3208 (2009-09-12)
Two approaches based on solidification of floating drop microextraction (SFDME) and homogenous liquid-liquid microextraction (HLLE) were compared for the extraction and preconcentration of di-(2-ethylhexyl) phthalate (DEHP) and di-(2-ethylhexyl) adipate (DEHA) from the mineral water samples. In SFDME, a floated drop

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service