Skip to Content
Merck
All Photos(1)

Key Documents

00310590

Quercetin 3-glucuronide

primary reference standard

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-glucuronide, Mikwelianin, Miquelianin, Quercetin 3-O-β-D-glucuronide, Querciturone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,000.00

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$1,000.00

About This Item

Empirical Formula (Hill Notation):
C21H18O13
CAS Number:
Molecular Weight:
478.36
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4

InChI

1S/C21H18O13/c22-7-4-10(25)12-11(5-7)32-17(6-1-2-8(23)9(24)3-6)18(13(12)26)33-21-16(29)14(27)15(28)19(34-21)20(30)31/h1-5,14-16,19,21-25,27-29H,(H,30,31)/t14-,15-,16+,19-,21+/m0/s1

InChI key

DUBCCGAQYVUYEU-ZUGPOPFOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: carum pimpinella

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service