Quercetin 3-glucuronide
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-glucuronide, Mikwelianin, Miquelianin, Quercetin 3-O-β-D-glucuronide, Querciturone
Empirical Formula (Hill Notation):
C21H18O13
CAS Number:
Molecular Weight:
478.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Research Area
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-3 of 3