Skip to Content
Merck
All Photos(1)

Key Documents

30-5020

Sigma-Aldrich

Tris Base

≥99.8%, solid

Synonym(s):

Tris Base, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
bp:
219-220 °C/10 mmHg (lit.)
Pricing and availability is not currently available.

Product Name

Tris(hydroxymethyl)aminomethane, ≥99.8%

Assay

≥99.8%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Suitability

for electrophoresis

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sylvain Clède et al.
Chemical communications (Cambridge, England), 48(62), 7729-7731 (2012-06-29)
A rhenium tris-carbonyl derivative has been designed to couple infrared and luminescent detection in cells. Both spectroscopies are consistent with one another; they point out the reliability of the present SCoMPI (for Single Core Multimodal Probe for Imaging) for bimodal
Corey A Baron et al.
Magnetic resonance in medicine, 73(3), 1075-1084 (2014-04-12)
An acquisition method that does not increase scan time or specific absorption rate is investigated for reducing the deleterious effects of cerebrospinal fluid (CSF) partial volume effects on diffusion tensor imaging (DTI) tractography. It is based on using a shorter
Filippo Bertoli et al.
Small (Weinheim an der Bergstrasse, Germany), 10(16), 3307-3315 (2014-04-17)
Nanoparticles in contact with cells and living organisms generate quite novel interactions at the interface between the nanoparticle surface and the surrounding biological environment. However, a detailed time resolved molecular level description of the evolving interactions as nanoparticles are internalized
D R Brocks et al.
Clinical pharmacokinetics, 23(6), 415-427 (1992-12-01)
Ketorolac is a new chiral nonsteroidal anti-inflammatory drug (NSAID) which is marketed for analgesia as the racemate. The drug is administered as the water soluble tromethamine salt and is available in tablets or as an intramuscular injection. The absorption of
Jihane Homman-Ludiye et al.
Cerebral cortex (New York, N.Y. : 1991), 24(11), 2884-2898 (2013-05-28)
The visual cortex is organized into discrete domains characterized by their specific function, connectivity, chemoarchitecture, and cytoarchitecture. Gradients of transcription factors across the anteroposterior and mediolateral axes of the neocortex have previously been demonstrated to specify the main sensory regions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service