Skip to Content
Merck
All Photos(3)

Key Documents

P5402

Sigma-Aldrich

Poly(ethylene glycol)

average mol wt 1,450

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$75.40
1 KG
$142.00
2.5 KG
$246.00

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$75.40
1 KG
$142.00
2.5 KG
$246.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder, flakes, or chunks

mol wt

average mol wt 1,450

pH

4.5-7.5

viscosity

26.5 cSt (210 °F)

mp

43-46 °C

solubility

water: soluble (approximately 720 mg/ml, 20 °C)

density

1.0919 g/mL at 60 °C
1.0761 g/mL at 80 °C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Composite scaffolds for bone regeneration: A study describes the development of a composite scaffold incorporating polyethylene glycol, gellan gum, and hydroxyapatite, with added ginseng derived Compound K, indicating potential applications in bone regeneration (Thangavelu et al., 2024).

  • Enhanced wound healing: Research discusses AI-driven optimization of PCL/PEG electrospun scaffolds, highlighting their improved efficacy in in vivo wound healing applications (Virijević et al., 2024).

  • Catch and release cryogels: Redox-responsive cryogels made from polyethylene glycol, which can capture and release proteins and cells, suggesting their potential in biomedical applications (Calik et al., 2024).

  • Antibacterial hydrogels for wound dressing: A study presents light-degradable nanocomposite hydrogels containing polyethylene glycol, aimed at antibacterial wound dressing applications, demonstrating innovative uses in medical materials (Fang et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benny Abraham Kaipparettu et al.
PloS one, 8(5), e61747-e61747 (2013-05-15)
Mitochondrial-nucleus cross talks and mitochondrial retrograde regulation can play a significant role in cellular properties. Transmitochondrial cybrid systems (cybrids) are an excellent tool to study specific effects of altered mitochondria under a defined nuclear background. The majority of the studies
Natalie Gavrilov-Yusim et al.
BMC biotechnology, 8, 3-3 (2008-01-16)
The isolation and production of human monoclonal antibodies is becoming an increasingly important pursuit as biopharmaceutical companies migrate their drug pipelines away from small organic molecules. As such, optimization of monoclonal antibody technologies is important, as this is becoming the
Corinne E Griguer et al.
PloS one, 3(11), e3655-e3655 (2008-11-06)
Mitochondria dysfunction and hypoxic microenvironment are hallmarks of cancer cell biology. Recently, many studies have focused on isolation of brain cancer stem cells using CD133 expression. In this study, we investigated whether CD133 expression is regulated by bioenergetic stresses affecting
Antonio Pantelias et al.
Cancers, 12(9) (2020-08-23)
For precision cancer radiotherapy, high linear energy transfer (LET) particle irradiation offers a substantial advantage over photon-based irradiation. In contrast to the sparse deposition of low-density energy by χ- or γ-rays, particle irradiation causes focal DNA damage through high-density energy
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service